Assay Details
| Gene Symbol: LOC4341800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788241.1 | XP_015643727.1 | 1 - 2 | 138 | 62 | |
| GenBank mRNA | AK073551.1 | - | 1 - 2 | 138 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788241.1 transcripts.
Set Membership: |
|

LOC4341800
Genomic Map