Assay Details
| Gene Symbol: LOC4341800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788240.2 | XP_015643726.1 | 9 - 10 | 828 | 68 | |
| XM_015788241.1 | XP_015643727.1 | 9 - 10 | 824 | 68 | ||
| XM_015788242.1 | XP_015643728.1 | 9 - 10 | 778 | 68 | ||
| XM_015788243.2 | XP_015643729.1 | 9 - 10 | 780 | 68 | ||
| GenBank mRNA | AK073551.1 | - | 9 - 10 | 824 | 68 | |
More Information
Set Membership: |
|

LOC4341800
Genomic Map