Assay Details
| Gene Symbol: LOC4341870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788190.2 | XP_015643676.1 | 1 - 2 | 167 | 67 | |
| XM_015788191.2 | XP_015643677.1 | 2 - 3 | 1653 | 67 | ||
| GenBank mRNA | AK068614.1 | - | 2 - 3 | 1659 | 67 | |
| AK120488.1 | - | 2 - 3 | 1659 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788191.2 transcripts.
Set Membership: |
|

LOC4341870
Genomic Map