Assay Details
| Gene Symbol: LOC4341870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788190.2 | XP_015643676.1 | 2 - 3 | 238 | 80 | |
| XM_015788191.2 | XP_015643677.1 | 3 - 4 | 1724 | 80 | ||
| GenBank mRNA | AK068614.1 | - | 3 - 4 | 1730 | 80 | |
| AK120488.1 | - | 3 - 4 | 1730 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015788191.2 transcripts.
Set Membership: |
|

LOC4341870
Genomic Map