Assay Details
| Gene Symbol: LOC4341870 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788190.2 | XP_015643676.1 | 6 - 7 | 519 | 146 | |
| XM_015788191.2 | XP_015643677.1 | 7 - 8 | 2005 | 146 | ||
| GenBank mRNA | AK068614.1 | - | 7 - 8 | 2011 | 146 | |
| AK120488.1 | - | 7 - 8 | 2011 | 146 | ||
More Information
Set Membership: |
|

LOC4341870
Genomic Map