Assay Details
| Gene Symbol: LOC4341876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785584.2 | XP_015641070.1 | 9 - 10 | 1988 | 57 | |
| XM_015785585.2 | XP_015641071.1 | 10 - 11 | 1855 | 57 | ||
| GenBank mRNA | AK102878.1 | - | 10 - 11 | 1784 | 57 | |
More Information
Set Membership: |
|

LOC4341876
Genomic Map