Assay Details
| Gene Symbol: LOC4341876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785584.2 | XP_015641070.1 | 1 - 2 | 415 | 93 | |
| XM_015785585.2 | XP_015641071.1 | 2 - 3 | 282 | 93 | ||
| GenBank mRNA | AK102878.1 | - | 2 - 3 | 211 | 93 | |
| AK103572.1 | - | 1 - 2 | 357 | 93 | ||
More Information
Set Membership: |
|

LOC4341876
Genomic Map