Assay Details
| Gene Symbol: LOC4341876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785584.2 | XP_015641070.1 | 3 - 4 | 901 | 154 | |
| XM_015785585.2 | XP_015641071.1 | 4 - 5 | 768 | 154 | ||
| GenBank mRNA | AK102878.1 | - | 4 - 5 | 697 | 154 | |
| AK103572.1 | - | 2 - 3 | 1034 | 154 | ||
More Information
Set Membership: |
|

LOC4341876
Genomic Map