Assay Details
| Gene Symbol: LOC4341876 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785584.2 | XP_015641070.1 | 7 - 8 | 1743 | 112 | |
| XM_015785585.2 | XP_015641071.1 | 8 - 9 | 1610 | 112 | ||
| GenBank mRNA | AK102878.1 | - | 8 - 9 | 1539 | 112 | |
| AK103572.1 | - | 5 - 6 | 2001 | 112 | ||
More Information
Set Membership: |
|

LOC4341876
Genomic Map