Assay Details
| Gene Symbol: LOC4341904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787901.2 | XP_015643387.1 | 10 - 11 | 1803 | 146 | |
| XM_015787902.2 | XP_015643388.1 | 10 - 11 | 1805 | 146 | ||
| XM_015787903.2 | XP_015643389.1 | 10 - 11 | 1805 | 146 | ||
| GenBank mRNA | AK068150.1 | - | 10 - 11 | 1795 | 146 | |
| AK100072.1 | - | 9 - 10 | 1378 | 146 | ||
More Information
Set Membership: |
|

LOC4341904
Genomic Map