Assay Details
| Gene Symbol: LOC4341904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787901.2 | XP_015643387.1 | 11 - 12 | 1904 | 138 | |
| XM_015787902.2 | XP_015643388.1 | 11 - 12 | 1906 | 138 | ||
| XM_015787903.2 | XP_015643389.1 | 11 - 12 | 1906 | 138 | ||
| GenBank mRNA | AK068150.1 | - | 11 - 12 | 1896 | 138 | |
| AK100072.1 | - | 10 - 11 | 1479 | 138 | ||
More Information
Set Membership: |
|

LOC4341904
Genomic Map