Assay Details
| Gene Symbol: LOC4341904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787901.2 | XP_015643387.1 | 15 - 16 | 2199 | 108 | |
| XM_015787902.2 | XP_015643388.1 | 15 - 16 | 2201 | 108 | ||
| XM_015787903.2 | XP_015643389.1 | 15 - 16 | 2201 | 108 | ||
| GenBank mRNA | AK068150.1 | - | 15 - 16 | 2191 | 108 | |
| AK100072.1 | - | 14 - 15 | 1774 | 108 | ||
More Information
Set Membership: |
|

LOC4341904
Genomic Map