Assay Details
| Gene Symbol: LOC4341904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787901.2 | XP_015643387.1 | 18 - 19 | 2747 | 106 | |
| GenBank mRNA | AK068150.1 | - | 18 - 19 | 2739 | 106 | |
| AK100072.1 | - | 17 - 18 | 2322 | 106 | ||
More Information
Set Membership: |
|

LOC4341904
Genomic Map