Assay Details
| Gene Symbol: LOC4341904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787901.2 | XP_015643387.1 | 6 - 7 | 1339 | 81 | |
| XM_015787902.2 | XP_015643388.1 | 6 - 7 | 1341 | 81 | ||
| XM_015787903.2 | XP_015643389.1 | 6 - 7 | 1341 | 81 | ||
| GenBank mRNA | AK068150.1 | - | 6 - 7 | 1331 | 81 | |
| AK100072.1 | - | 5 - 6 | 914 | 81 | ||
More Information
Set Membership: |
|

LOC4341904
Genomic Map