Assay Details
| Gene Symbol: LOC4341904 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787901.2 | XP_015643387.1 | 9 - 10 | 1679 | 84 | |
| XM_015787902.2 | XP_015643388.1 | 9 - 10 | 1681 | 84 | ||
| XM_015787903.2 | XP_015643389.1 | 9 - 10 | 1681 | 84 | ||
| GenBank mRNA | AK100072.1 | - | 8 - 9 | 1254 | 84 | |
More Information
Set Membership: |
|

LOC4341904
Genomic Map