Assay Details
| Gene Symbol: LOC4342017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787582.1 | XP_015643068.1 | 5 - 6 | 1267 | 75 | |
| GenBank mRNA | AK066531.1 | - | 5 - 6 | 1204 | 75 | |
| AK067925.1 | - | 4 - 5 | 1434 | 75 | ||
More Information
Set Membership: |
|

LOC4342017
Genomic Map