Assay Details
| Gene Symbol: LOC4342017 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787582.1 | XP_015643068.1 | 7 - 8 | 1556 | 74 | |
| GenBank mRNA | AK066531.1 | - | 7 - 8 | 1493 | 74 | |
| AK067925.1 | - | 6 - 7 | 1723 | 74 | ||
| AK070823.1 | - | 2 - 3 | 162 | 74 | ||
More Information
Set Membership: |
|

LOC4342017
Genomic Map