Assay Details
| Gene Symbol: LOC4342124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787591.2 | XP_015643077.1 | 1 - 2 | 215 | 78 | |
| GenBank mRNA | AK069446.1 | - | 1 - 2 | 123 | 78 | |
| AK100019.1 | - | 1 - 2 | 211 | 78 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015787591.2 transcripts.
Set Membership: |
|

LOC4342124
Genomic Map