Assay Details
| Gene Symbol: LOC4342124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787591.2 | XP_015643077.1 | 4 - 5 | 1363 | 77 | |
| GenBank mRNA | AK069446.1 | - | 4 - 5 | 1271 | 77 | |
| AK100019.1 | - | 4 - 5 | 1359 | 77 | ||
| D26484.1 | - | 4 - 5 | 1296 | 77 | ||
More Information
Set Membership: |
|

LOC4342124
Genomic Map