Assay Details
| Gene Symbol: LOC4342124 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015787591.2 | XP_015643077.1 | 5 - 6 | 1459 | 109 | |
| GenBank mRNA | AK069446.1 | - | 5 - 6 | 1367 | 109 | |
| AK100019.1 | - | 5 - 6 | 1455 | 109 | ||
| D26484.1 | - | 5 - 6 | 1392 | 109 | ||
More Information
Set Membership: |
|

LOC4342124
Genomic Map