Assay Details
| Gene Symbol: LOC4342377 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790081.2 | XP_015645567.1 | 1 | 739 | 93 | |
| GenBank mRNA | AK108823.1 | - | 1 | 745 | 93 | |
More Information
Set Membership: |
|

LOC4342377
Genomic Map