Assay Details
| Gene Symbol: LOC4342738 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790669.2 | XP_015646155.1 | 1 | 619 | 121 | |
| GenBank mRNA | AK107870.1 | - | 1 | 548 | 121 | |
More Information
Set Membership: |
|

LOC4342738
Genomic Map