Assay Details
| Gene Symbol: LOC4343074 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792076.2 | XP_015647562.1 | 1 - 2 | 2467 | 69 | |
| GenBank mRNA | AK067172.1 | - | 1 - 2 | 2468 | 69 | |
More Information
Set Membership: |
|

LOC4343074
Genomic Map