Assay Details
| Gene Symbol: LOC4343198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791878.2 | XP_015647364.1 | 6 - 7 | 2787 | 126 | |
| XM_015791879.2 | XP_015647365.1 | 5 - 6 | 2189 | 126 | ||
| GenBank mRNA | AK068617.1 | - | 6 - 7 | 2793 | 126 | |
More Information
Set Membership: |
|

LOC4343198
Genomic Map