Assay Details
| Gene Symbol: LOC4343265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790945.2 | XP_015646431.1 | 3 - 4 | 4704 | 69 | |
| XM_015790946.2 | XP_015646432.1 | 4 - 5 | 528 | 69 | ||
| GenBank mRNA | AK070976.1 | - | 4 - 5 | 453 | 69 | |
More Information
Set Membership: |
|

LOC4343265
Genomic Map