Assay Details
| Gene Symbol: LOC4343265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790945.2 | XP_015646431.1 | 5 - 6 | 4911 | 153 | |
| XM_015790946.2 | XP_015646432.1 | 6 - 7 | 735 | 153 | ||
| GenBank mRNA | AK070976.1 | - | 6 - 7 | 660 | 153 | |
More Information
Set Membership: |
|

LOC4343265
Genomic Map