Assay Details
| Gene Symbol: LOC4343301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791591.2 | XP_015647077.1 | 13 - 14 | 2144 | 73 | |
| XM_015791592.2 | XP_015647078.1 | 12 - 13 | 1972 | 73 | ||
| GenBank mRNA | AK068275.1 | - | 13 - 14 | 1604 | 73 | |
More Information
Set Membership: |
|

LOC4343301
Genomic Map