Assay Details
| Gene Symbol: LOC4343301 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791592.2 | XP_015647078.1 | 2 - 3 | 498 | 107 | |
| GenBank mRNA | AK068275.1 | - | 2 - 3 | 468 | 107 | |
More Information
Set Membership: |
|

LOC4343301
Genomic Map