Assay Details
| Gene Symbol: LOC4352284 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK242914.1 | - | 3858 | 110 | ||
| Gene Symbol: LOC4343319 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790308.2 | XP_015645794.1 | 1 | 762 | 110 | |
More Information
Set Membership: |
|

LOC4352284
Genomic Map