Assay Details
| Gene Symbol: LOC4343570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791652.2 | XP_015647138.1 | 1 - 2 | 161 | 125 | |
| GenBank mRNA | AF093631.1 | - | 1 - 2 | 85 | 125 | |
| AK071634.1 | - | 1 - 2 | 97 | 125 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791652.2 transcripts.
Set Membership: |
|

LOC4343570
Genomic Map