Assay Details
| Gene Symbol: LOC4343570 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791652.2 | XP_015647138.1 | 2 - 3 | 396 | 80 | |
| GenBank mRNA | AF093631.1 | - | 2 - 3 | 320 | 80 | |
| AK071634.1 | - | 2 - 3 | 332 | 80 | ||
| Gene Symbol: LOC9271026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK241277.1 | - | 1395 | 80 | ||
More Information
Set Membership: |
|

LOC4343570
Genomic Map