Assay Details
| Gene Symbol: LOC4343735 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791259.2 | XP_015646745.1 | 1 - 2 | 179 | 147 | |
| GenBank mRNA | AK073470.1 | - | 1 - 2 | 137 | 147 | |
| AK098890.1 | - | 1 - 2 | 137 | 147 | ||
| AK122131.1 | - | 1 - 2 | 185 | 147 | ||
| AY062210.2 | - | 1 - 2 | 125 | 148 | ||
| Gene Symbol: LOC4325515 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK287679.1 | - | 3262 | 147 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015791259.2 transcripts.
Set Membership: |
|

LOC4343735
Genomic Map