Assay Details
| Gene Symbol: LOC4343841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792364.1 | XP_015647850.1 | 3 - 4 | 609 | 109 | |
| XM_015792365.2 | XP_015647851.1 | 2 - 3 | 283 | 109 | ||
| GenBank mRNA | AK101099.1 | - | 3 - 4 | 495 | 109 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792365.2 transcripts.
Set Membership: |
|

LOC4343841
Genomic Map