Assay Details
| Gene Symbol: LOC4343841 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792364.1 | XP_015647850.1 | 5 - 6 | 1371 | 76 | |
| XM_015792365.2 | XP_015647851.1 | 4 - 5 | 1045 | 76 | ||
| XM_015792367.2 | XP_015647853.1 | 3 - 4 | 684 | 76 | ||
| GenBank mRNA | AK061822.1 | - | 1 - 2 | 50 | 76 | |
| AK101099.1 | - | 5 - 6 | 1257 | 76 | ||
More Information
Set Membership: |
|

LOC4343841
Genomic Map