Assay Details
| Gene Symbol: LOC4343962 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790192.2 | XP_015645678.1 | 1 | 577 | 72 | |
| GenBank mRNA | AK073278.1 | - | 1 | 454 | 72 | |
More Information
Set Membership: |
|

LOC4343962
Genomic Map