Assay Details
| Gene Symbol: LOC4344009 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791622.2 | XP_015647108.1 | 2 - 3 | 308 | 81 | |
| XM_015791623.2 | XP_015647109.1 | 2 - 3 | 308 | 81 | ||
| GenBank mRNA | AK061338.1 | - | 2 - 3 | 260 | 81 | |
| AK073451.1 | - | 2 - 3 | 312 | 81 | ||
| AK287924.1 | - | 1415 | 81 | |||
More Information
Set Membership: |
|

LOC4344009
Genomic Map