Assay Details
| Gene Symbol: LOC4344041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790576.2 | XP_015646062.1 | 5 - 6 | 1065 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC4344041| Gene Symbol: LOC4344041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790576.2 | XP_015646062.1 | 5 - 6 | 1065 | 127 | |
Set Membership: |
|