Assay Details
| Gene Symbol: LOC4344085 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789594.1 | XP_015645080.1 | 3 - 4 | 871 | 87 | |
| GenBank mRNA | AK064653.1 | - | 3 - 4 | 856 | 87 | |
More Information
Set Membership: |
|

LOC4344085
Genomic Map