Assay Details
| Gene Symbol: LOC9270916 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789578.2 | XP_015645064.1 | 3 | 1499 | 93 | |
| GenBank mRNA | AK241936.1 | - | 1 | 578 | 93 | |
More Information
Set Membership: |
|

LOC9270916
Genomic Map