Assay Details
| Gene Symbol: LOC4344248 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792200.2 | XP_015647686.1 | 12 - 13 | 1370 | 101 | |
| XM_015792201.2 | XP_015647687.1 | 12 - 13 | 1370 | 101 | ||
| GenBank mRNA | AK065980.1 | - | 12 - 13 | 1361 | 101 | |
More Information
Set Membership: |
|

LOC4344248
Genomic Map