Assay Details
| Gene Symbol: LOC4344493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795305.2 | XP_015650791.1 | 3 - 4 | 750 | 77 | |
| GenBank mRNA | AB003326.1 | - | 3 - 4 | 548 | 77 | |
| AK069122.1 | - | 3 - 4 | 580 | 77 | ||
| AY551925.1 | - | 3 - 4 | 439 | 77 | ||
More Information
Set Membership: |
|

LOC4344493
Genomic Map