Assay Details
| Gene Symbol: LOC4344493 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795305.2 | XP_015650791.1 | 4 - 5 | 854 | 96 | |
| GenBank mRNA | AB003326.1 | - | 4 - 5 | 652 | 96 | |
| AK069122.1 | - | 4 - 5 | 684 | 96 | ||
| AY551925.1 | - | 4 - 5 | 543 | 96 | ||
More Information
Set Membership: |
|

LOC4344493
Genomic Map