Assay Details
| Gene Symbol: LOC4344521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794374.2 | XP_015649860.1 | 11 - 12 | 1113 | 183 | |
| XM_015794376.2 | XP_015649862.1 | 11 - 12 | 1160 | 183 | ||
| XM_015794377.2 | XP_015649863.1 | 11 - 12 | 941 | 183 | ||
| XM_015794378.2 | XP_015649864.1 | 11 - 12 | 844 | 183 | ||
| XM_015794379.2 | XP_015649865.1 | 11 - 12 | 1122 | 183 | ||
| XM_015794382.2 | XP_015649868.1 | 11 - 12 | 806 | 183 | ||
| XM_015794383.2 | XP_015649869.1 | 11 - 12 | 1073 | 183 | ||
| XM_015794384.2 | XP_015649870.1 | 9 - 10 | 610 | 183 | ||
| XM_015794385.2 | XP_015649871.1 | 9 - 10 | 550 | 183 | ||
| XM_015794386.2 | XP_015649872.1 | 9 - 10 | 640 | 183 | ||
| XM_015794387.2 | XP_015649873.1 | 9 - 10 | 653 | 183 | ||
| GenBank mRNA | AK287457.1 | - | 11 - 12 | 948 | 183 | |
More Information
Set Membership: |
|

LOC4344521
Genomic Map