Assay Details
| Gene Symbol: LOC4344521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794374.2 | XP_015649860.1 | 4 - 5 | 703 | 81 | |
| XM_015794376.2 | XP_015649862.1 | 4 - 5 | 750 | 81 | ||
| XM_015794377.2 | XP_015649863.1 | 4 - 5 | 531 | 81 | ||
| XM_015794378.2 | XP_015649864.1 | 4 - 5 | 434 | 81 | ||
| XM_015794379.2 | XP_015649865.1 | 4 - 5 | 712 | 81 | ||
| XM_015794382.2 | XP_015649868.1 | 4 - 5 | 396 | 81 | ||
| XM_015794383.2 | XP_015649869.1 | 4 - 5 | 663 | 81 | ||
| XM_015794384.2 | XP_015649870.1 | 2 - 3 | 200 | 81 | ||
| GenBank mRNA | AK287457.1 | - | 4 - 5 | 538 | 81 | |
More Information
Set Membership: |
|

LOC4344521
Genomic Map