Assay Details
| Gene Symbol: LOC4344521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794374.2 | XP_015649860.1 | 5 - 6 | 727 | 69 | |
| XM_015794376.2 | XP_015649862.1 | 5 - 6 | 774 | 69 | ||
| XM_015794377.2 | XP_015649863.1 | 5 - 6 | 555 | 69 | ||
| XM_015794378.2 | XP_015649864.1 | 5 - 6 | 458 | 69 | ||
| XM_015794379.2 | XP_015649865.1 | 5 - 6 | 736 | 69 | ||
| XM_015794382.2 | XP_015649868.1 | 5 - 6 | 420 | 69 | ||
| XM_015794383.2 | XP_015649869.1 | 5 - 6 | 687 | 69 | ||
| XM_015794384.2 | XP_015649870.1 | 3 - 4 | 224 | 69 | ||
| XM_015794385.2 | XP_015649871.1 | 3 - 4 | 164 | 69 | ||
| XM_015794386.2 | XP_015649872.1 | 3 - 4 | 254 | 69 | ||
| XM_015794387.2 | XP_015649873.1 | 3 - 4 | 267 | 69 | ||
| GenBank mRNA | AK287457.1 | - | 5 - 6 | 562 | 69 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015794386.2, XM_015794387.2 transcripts.
Set Membership: |
|

LOC4344521
Genomic Map