Assay Details
| Gene Symbol: LOC4344521 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794374.2 | XP_015649860.1 | 7 - 8 | 871 | 85 | |
| XM_015794376.2 | XP_015649862.1 | 7 - 8 | 918 | 85 | ||
| XM_015794377.2 | XP_015649863.1 | 7 - 8 | 699 | 85 | ||
| XM_015794378.2 | XP_015649864.1 | 7 - 8 | 602 | 85 | ||
| XM_015794379.2 | XP_015649865.1 | 7 - 8 | 880 | 85 | ||
| XM_015794382.2 | XP_015649868.1 | 7 - 8 | 564 | 85 | ||
| XM_015794383.2 | XP_015649869.1 | 7 - 8 | 831 | 85 | ||
| XM_015794384.2 | XP_015649870.1 | 5 - 6 | 368 | 85 | ||
| XM_015794385.2 | XP_015649871.1 | 5 - 6 | 308 | 85 | ||
| XM_015794386.2 | XP_015649872.1 | 5 - 6 | 398 | 85 | ||
| XM_015794387.2 | XP_015649873.1 | 5 - 6 | 411 | 85 | ||
| GenBank mRNA | AK287457.1 | - | 7 - 8 | 706 | 85 | |
More Information
Set Membership: |
|

LOC4344521
Genomic Map