Assay Details
| Gene Symbol: LOC4344545 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015794216.2 | XP_015649702.1 | 1 - 2 | 231 | 85 | |
| GenBank mRNA | AK069887.1 | - | 1 - 2 | 129 | 85 | |
| AK104952.1 | - | 1 - 2 | 128 | 85 | ||
More Information
Set Membership: |
|

LOC4344545
Genomic Map