Assay Details
| Gene Symbol: LOC4345056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795237.2 | XP_015650723.1 | 13 - 14 | 2435 | 87 | |
| GenBank mRNA | AK100945.1 | - | 13 - 14 | 2383 | 87 | |
More Information
Set Membership: |
|

LOC4345056
Genomic Map