Assay Details
| Gene Symbol: LOC4345056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795237.2 | XP_015650723.1 | 5 - 6 | 1418 | 137 | |
| GenBank mRNA | AK100945.1 | - | 5 - 6 | 1366 | 137 | |
More Information
Set Membership: |
|

LOC4345056
Genomic Map