Assay Details
| Gene Symbol: LOC4345056 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015795237.2 | XP_015650723.1 | 6 - 7 | 1562 | 99 | |
| GenBank mRNA | AK100945.1 | - | 6 - 7 | 1510 | 99 | |
More Information
Set Membership: |
|

LOC4345056
Genomic Map